Trigger Bottles, Sterile, Veltek
Supplier: Veltek Associates
Certificates
About this item
Ideal for dispensing disinfecting agents, sporicidal agents, and other solutions.
Bottles are washed, gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation.