ContactUsLinkComponent
Barnstead™ Single Filter Holder, Economy, Thermo Scientific
  26303-020
undefined
Barnstead™ Single Filter Holder, Economy, Thermo Scientific
  26303-020
 :  15840
  • Description:
    Economy Filter Holder
  • Max. Inlet Pressure:
    100 psi
  • Cat. No.:
    26303-020
  • Supplier no.:
    15840
Single filter holder accepts Thermo Scientific double open-end prefilters or final filters.
  • Reinforced PP construction
  • Features push-button air bleed and Buna-N, O-rings
  • Inlet/Outlet connections are 3/4" NPT