Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Sample Tubes, PFA, BrandTech

Sample Tubes, PFA, BrandTech

15 ml (0.5 oz.), 22dia. x 110H mm (7/8 x 45/16")
Catalog # 83008-562
Supplier:   Brandtech
Supplier #:  103897
Sample Tubes, PFA, BrandTech
Catalog # 83008-562
Supplier:  Brandtech
Sample Tubes, PFA, BrandTech
Sample Tubes, PFA, BrandTech

15 ml (0.5 oz.), 22dia. x 110H mm (7/8 x 45/16")

Price
$285.40
Log in for availability
Pack of 1
Minimum order quantity is 1
Product Details & Documents

Sample tubes used for sample preparation, centrifugation, and autosampler racks for trace analysis.

  • Made of PFA (perfluoroalkoxy copolymer)
  • Highly heat- and chemical-resistant, autoclavable, and easy-to-clean
  • Can withstand temperatures from –200 to 260 °C (–328 to 500 °F)

The 15mL sample tube features a calibrated ring-mark at 10mL and a PFA screw cap with a GL25 screw thread.

Specifications
  • Capacity:
    15 ml (0.5 oz.)
  • Dimensions:
    22dia. x 110H mm (7/8 x 45/16")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.