Aerosolve® Canisters for SX4750, SX4750A, and JS-4.3 Rotors, Beckman Coulter
These canisters for use with the SX4750 and SX4750A rotors (BK369702 series) and JS-4.3 (BK362734) swinging bucket rotor.
- Clear plastic for viewing top to bottom
- O-ring seals
- Autoclavable up to 121 °C (249 °F)
- Usable as a 500 ml wide-mouth bottle
Canisters help minimize contamination from broken or leaking tubes.
Note: Prior to opening, the sealed canister should be examined for breakage.
: Each Aerosolve® canister is supplied with base, cap, O-ring, and pad (also available separately). Tube racks, which are required for use with canisters, are sold separately.
Frequently Bought Together