ContactUsLinkComponent
Tube Decanter for 10/12 mm Modular Disk Adapters, High Performance Rotors, Beckman Coulter®
Tube Decanter for 10/12 mm Modular Disk Adapters, High Performance Rotors, Beckman Coulter®
  BK343108
undefined
Tube Decanter for 10/12 mm Modular Disk Adapters, High Performance Rotors, Beckman Coulter®
  BK343108
 :  343108
  • Description:
    Tube Decanter for 12 mm Modular Disk Adapters
  • Cat. No.:
    BK343108
  • Supplier no.:
    343108
For decanting a full rack of RIA tubes at once. Includes silicone gaskets for 10- and 12-mm tubes.
For use in blue modular disk adapters (part no. BK356148, BK359469).
For use in High Performance and High Capacity Rotors: JS-3.0 (BK339081) JS-5.2 (BK339087) JS-4.0 (BK339086) JS-4.2 (BK339080) JS-4.2A (BK366695).