Order Entry
United States
ContactUsLinkComponent
Accessories for MetroMax™ Mobile Desk, Metro International
Accessories for MetroMax™ Mobile Desk, Metro International
Catalog # 82000-932
Supplier:  Metro
undefined
Accessories for MetroMax™ Mobile Desk, Metro International
Catalog # 82000-932
Supplier:  Metro
Supplier Number:  FMBX

Specifications

  • Width:
    3⁷⁄₁₆"
  • Description:
    Flat Monitor Mounting Bracket
  • Height:
    5¾"
  • Length:
    8¹³⁄₁₆"
  • Environmentally Preferable:
  • Cat. No.:
    82000-932
  • Shipping weight:
    4 lbs.
  • Load capacity:
    30 lbs.
  • Supplier no.:
    FMBX

Specifications

About this item