About this item
Standards are packaged in acetone at a concentration of 200 µg/mL in ampoules.
- 200 μg/mL in acetone, 1 mL/ampoule
- Mixture contains 2,4,5,6-Tetrachloro-m-Xylene (TCMX) and Decachlorobiphenyl (DCB)
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- CAS No.:877-09-8
- Formula:C₈H₆Cl₄
- Description:2,4,5,6-Tetrachloro-m-Xylene