ContactUsLinkComponent
UST Diesel Fuel No. 2 Composite Standard, Restek
UST Diesel Fuel No. 2 Composite Standard, Restek
Catalog # RK31093
Supplier:  Restek
CAS Number:  
undefined
UST Diesel Fuel No. 2 Composite Standard, Restek
Catalog # RK31093
Supplier:  Restek
Supplier Number:  31093
CAS Number:  
Specifications
  • Description:
    Diesel Fuel No. 2 Composite Standard
  • Cat. No.:
    RK31093
  • Supplier no.:
    31093
Specifications
About this item
Standard is packaged in methylene chloride at a concentration of 5000 µg/mL in a 1 mL ampoule.
  • Diesel fuel standards may exhibit lot-to-lot variation
  • 5000 μg/mL in methylene chloride, 1 mL/ampoule
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).