EPA Method 501 Trihalomethane Mixture, Ultra Scientific
Catalog # ULTHM-521-1
Supplier: Ultra Scientific
Small Business Enterprise
CAS Number:
Specifications
- Concentration:5000 µg/ml
- Cat. No.:ULTHM-521-1
- No. of analytes:4
- Supplier no.:THM-521-1
Specifications
About this item
Mixture is packaged in methyl alcohol in 1 mL ampules.