ContactUsLinkComponent
LUST Retention Time Standard, Restek
Catalog # RK31200
Supplier:  Restek
CAS Number:  
undefined
LUST Retention Time Standard, Restek
Catalog # RK31200
Supplier:  Restek
Supplier Number:  31200
CAS Number:  
Specifications
  • Description:
    LUST Retention Time Standard
  • Cat. No.:
    RK31200
  • Supplier no.:
    31200
Specifications
About this item
Leaking Underground Storage Tank (LUST) standards are packaged in methylene chloride at a concentration of 25 µg/mL in a 1 mL ampoule.
  • 25 µg/mL each in methylene chloride, 1 mL/ampoule
  • Contains decane (C10), dodecane (C12), hexane (C6), hectacosane (C28), tetracontane (C40), tetracosane (C24), and triacontane (C30)
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).