ContactUsLinkComponent
EPA method 604 Phenols Calibration Mix, Restek
EPA method 604 Phenols Calibration Mix, Restek
Catalog # RK31029
Supplier:  Restek
CAS Number:  
undefined
EPA method 604 Phenols Calibration Mix, Restek
Catalog # RK31029
Supplier:  Restek
Supplier Number:  31029
CAS Number:  
Specifications
  • Description:
    EPA Method 604 Phenols Calibration Mix
  • Cat. No.:
    RK31029
  • Supplier no.:
    31029
Specifications
About this item
Mixture is packaged in methanol at a concentration of 2000 µg/mL in a 1 mL ampoule.
  • Mixture contains 4-Chloro-3-methylphenol, 2-Chlorophenol, 2,4-Dichlorophenol, 2,4-Dimethylphenol, 2,4-Dinitrophenol, 2-Methyl-4,6-dinitrophenol, 2-Nitrophenol, 4-Nitrophenol, Pentachlorophenol, Phenol, and 2,4,6-Trichlorophenol
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).