ContactUsLinkComponent
EPA Method 8270 and CLP–Semi-Volatiles (3/90 SOW) Internal Standards, NSI
EPA Method 8270 and CLP–Semi-Volatiles (3/90 SOW) Internal Standards, NSI
Catalog # NSC-394H
Supplier:  SPEX CERTIPREP LLC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
EPA Method 8270 and CLP–Semi-Volatiles (3/90 SOW) Internal Standards, NSI
Catalog # NSC-394H
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  C-394H
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    EPA Method 8270 and CLP–Semi-Volatiles (3/90 SOW) Internal Standards
  • Cat. No.:
    NSC-394H
  • Supplier no.:
    C-394H
Specifications
About this item
Mixture is packaged in 4:1 methylene chloride/carbon disulfide at a concentration of 4000µg/ml in methylene chloride/carbon disulfide (4:1) in a 1.5ml ampule
Mixture contains Acenaphthene-d10, Chrysene-d12, 1,4-Dichlorobenzene-d4, Naphthalene-d8, Perylene-d12, and Phenanthrene-d10.
An A2LA traceable standard that has been second source verified in house using an A2LA verified standard.