Low-Level Solids Wastewater Standard, NSI
Catalog # NS-QCI-057
Supplier: SPEX CERTIPREP LLC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Ready-To-Use
- Size:4×1L
- Cat. No.:NS-QCI-057
- Supplier no.:QCI-057
Specifications
About this item
Certified for TSS
Standard contains 15–30 mg/L of TSS.
Includes CoA listing true value and acceptance limit.