Complex Nutrients Standard, NSI
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Ready-to-use whole volume standard
Standard contains 1.50–35 mg/L of TKN and 0.50–10 mg/L of Total Phosphorus.
Includes CoA listing true value and acceptance limits. Supplied in HDPE bottle.
Specifications
Specifications
- Size:500 ml