Order Entry
Puerto Rico
ContactUsLinkComponent
 
Multidrop® Combi Reagent Dispenser, Thermo Scientific
  11388-558
 :  Avantor
undefined
Multidrop® Combi Reagent Dispenser, Thermo Scientific
  11388-558
 :  Avantor

 

  • Description:
    Multidrop® Combi Dispenser
  • Electrical:
    100–240V, 50/60Hz
  • Dimensions:
    35.5W x 33D x 22H cm (14 x 13 x 811/16")
  • Weight:
    9.1 kg (20 lbs.)
  • Cat. No.:
    11388-558
  • Supplier no.:
    5840300

 

 

Ideal for demanding pharmaceutical and biotech laboratories, this high-speed dispenser is compatible with 6- to 1536-well plates with a height of 5–50mm. Dispenser precisely dispenses volumes of 0.5–2500µL in 0.5 and 5µL increments. Dispensing a total of 20µL per plate takes only four seconds for a 96-well plate and nine seconds for a 384-well plate. Dispensing a total of 1µL to a 1536-well plate takes only 14 seconds. Precision CV is ≤1.5% at 20µL and it is typically accurate to ±2% at 20µL.

Multidrop Combi SMART option (89037-720) adds an advanced built-in tracking system for improved reliability and cassette lifetime traceability, as well as greatly enhancing efficiency and reporting capability.

Multidrop® Combi SMART option (89037-720) adds an advanced built-in tracking system for improved reliability and cassette lifetime traceability, as well as greatly enhancing efficiency and reporting capability.

Small tube dispensing cassettes are to be used with volumes from 0.5–50µL. Standard tube dispensing cassettes are for volumes from 5–2500µL.

Dispenser precisely dispenses volumes of 0.5–2500µL in 0.5 and 5µL increments. Dispensing a total of 20µL per plate takes only four seconds for a 96-well plate and nine seconds for a 384-well plate. Dispensing a total of 1µL to a 1536-well plate takes only 14 seconds. Precision CV is ≤1.5% at 20µL and it is typically accurate to ±2% at 20µL.

 : Dispenser 11388-558 includes dispensing cassettes 14217-954, 14230-390, and -388. Dispenser 89037-720 includes dispensing cassettes 89037-722, -726, and -730. Factory developed installation, operation, and performance qualification document is approved by design engineers and written in accordance with standards and guidelines. The qualification services are carried out by certified technicians on site.