ContactUsLinkComponent
Synthware Stopcock, 1:5 PTFE Plug, Metering Valve, Kemtech America
Synthware Stopcock, 1:5 PTFE Plug, Metering Valve, Kemtech America
Catalog # 60005-126
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Stopcock, 1:5 PTFE Plug, Metering Valve, Kemtech America
Catalog # 60005-126
Supplier:  Kemtech America
Supplier Number:  S191530
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Description:
    Plug
  • Material:
    PTFE
  • Cat. No.:
    60005-126
  • Stopcock size:
    4 mm
  • Supplier no.:
    S191530
Specifications
About this item
Replacement Teflon® metering stopcock.
  • Teflon® construction
  • Metering valve
  • Size 15
With a threaded Teflon® needle valve for the fine control of flowthrough the bore of the stopcock plug. This replacement kit is complete with all the components.