Synthware Vacuum Manifold with PTFE Stopcocks, Kemtech America
Supplier: Kemtech America
Certificates
About this item
General purpose manifolds are available with either 2 mm or 4 mm PTFE stopcocks
- Heavy wall borosilicate construction
- Teflon® stopcocks
- 4 or 6 port options available
The main tube has a 22 mm O.D., and features a serrated hose connection (10 mm O.D.) at the right, left, or both right and left sides. Delivery ports have 10 mm serrated hose connections. Distance between stopcocks is 100 mm.