To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Max. Sample Capacity:160 Samples
- Unit Dimensions:39.4W x 25.4D x 8.9H cm (151/2 x 10 x 31/2")
- Description:HR2025 Horizontal Gel Electrophoresis System
- Environmentally Preferable:
- Gel Dimensions:20W x 25L cm (77/8 x 97/8")
- Cat. No.:IB56000
- Buffer volume:1.2 L
- Supplier no.:56000
Ideal for inversion and genomic DNA separations
Electrical gravity connector minimizes electrical hazards and eliminates sample disturbance during setup. Gold-plated electrical fittings and 99.9% platinum electrodes resist corrosion and withstand rapid, high voltage polarity switching applications.
Buffer recirculation capability permits high resolution electrophoresis of nucleic acids in low ionic strength buffers. Dedicated casting system eliminates the need for tape and permits gel preparation while minimizing leaks. Well visualization stripes improve visibility to facilitate gel loading, even with a sample quick loader. Vented lid reduces condensation, improving resolution of runs. UVT gel tray with multiple comb slots allows maximum sample capacity using several combs, or maximum running lengths using a single comb. Tray transmits 312nm UV light.