Soldering Tips for Aven Etneo™ SX and Weller® Stations, Plato™, ITW Chemtronics
Supplier: ITW Chemtronics
Certificates
About this item
The interior surfaces of these tips are protected to prevent adhesion to the sensor.
Tips can be used with Aven Etneo™ SX stations and Weller® stations EC-1002, EC-2002, MC-5001, WCC-100, WTL-1000, DTL-1000, and DEC-1000, which are used with EC1201A, EC1201AFE, EX1201AFE, and WCC-101 soldering irons.
Certifications: Tips meet or exceed OEM specifications.
Frequently Bought Together