Order Entry
Puerto Rico
ContactUsLinkComponent
Airfuge® Ultracentrifuges, Beckman Coulter®
Airfuge® Ultracentrifuges, Beckman Coulter®
Catalog # BK340400
Supplier:  Beckman Coulter
Product available on GSA Advantage®
undefined
Airfuge® Ultracentrifuges, Beckman Coulter®
Catalog # BK340400
Supplier:  Beckman Coulter
Supplier Number:  340400
Product available on GSA Advantage®

Specifications

  • Description:
    Airfuge® Ultracentrifuge
  • Shipping Weight:
    15.9 kg (35 lbs.)
  • Pressure:
    30 psig (210 kPa) for routine operation
  • Acceleration/Braking Time:
    30 - 60 s / 2.5–5.5 min.
  • Dimensions:
    28W x 20H x 38L cm (11 x 8 x 15")
  • Cat. No.:
    BK340400
  • Electrical:
    120 V, 60 Hz
  • Max. speed:
    110,000 rpm
  • Max. rcf:
    199000×g
  • Supplier no.:
    340400

Specifications

About this item

This compact ultracentrifuge is a versatile and convenient benchtop unit. A variety of rotors make the Airfuge ideal for applications such as receptor-binding studies, lipoprotein assays, chylomicron studies, sample preparation for electron microscopy, and clarification of lipemic samples.

  • Versatile and Convenient Benchtop Unit
  • Air Turbine-Driven
  • Results within Short Run Times
  • Ideal for Receptor-Binding Studies and Lipoprotein Assays
  • Variety of Rotors

The Airfuge® is air turbine driven and delivers results in a fraction of the time of larger units

The speed is air-pressure regulated and braking is first achieved by coasting, then friction braking by engagement of the brake pin. The rotor operates at a temperature a few degrees above ambient.

The combination of high g-force and short path length results in remarkably short run times to produce results. Runs can be timed from 0–300 minutes in 5 minute increments; a HOLD button can be used for longer runs.

Ordering information: Supplied with a manufacturer's one-year instrument and rotor warranty. Complete with connecting hose and air filter. Rotors sold separately (BK347593 series).