Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
PYREX® Tissue Grinders, PTFE pestle, Corning

PYREX® Tissue Grinders, PTFE pestle, Corning

1 ml, 13×110 mm
Catalog # 22877-240
Supplier:   Corning
Supplier #:  7724T-1
PYREX® Tissue Grinders, PTFE pestle, Corning
Catalog # 22877-240
Supplier:  Corning
PYREX® Tissue Grinders, PTFE pestle, Corning
PYREX® Tissue Grinders, PTFE pestle, Corning

1 ml, 13×110 mm

Price
$382.00
Log in for availability
Case of 1
Product Details & Documents

Homogenizers designed for soft tissue found in liver or brain. They perform initial grinding and final homogenation in same sequence. Glass pestle has fluoropolymer resin contact surface threaded onto a stainless steel shaft. Mortar is not ground. All tissue grinders have a pourout top.

Caution: Caution: If used with motor-driven device, attachment must contain some form of friction release clutch to prevent connective tissue or muscle from damaging apparatus. User should wear protective gloves of sufficient thickness in case glass components fail.

Documents

Specifications
  • Capacity:
    1 ml
  • O.D.×H:
    13×110 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.