Order Entry
Puerto Rico
ContactUsLinkComponent
PRECISION™ Disposable Tissue Grinder System, Covidien
PRECISION™ Disposable Tissue Grinder System, Covidien
Catalog # 15704-124
Supplier:  Cardinal Health
undefined
PRECISION™ Disposable Tissue Grinder System, Covidien
Catalog # 15704-124
Supplier:  Cardinal Health
Supplier Number:  3500SA

Specifications

  • Capacity:
    50 ml
  • Description:
    Disposable Tissue Grinder System
  • Cat. No.:
    15704-124
  • Supplier no.:
    3500SA

Specifications

About this item

Grinder comes with a 50mL tube that can double as a specimen container for transport and storage

A protective sleeve helps contain any aerosolization created during grinding. Design allows specimen to be cooled while grinding. Supplied in a sterile pouch.