ContactUsLinkComponent
Disposable Tissue Grinder System, Covidien
Disposable Tissue Grinder System, Covidien
Catalog # 15704-126
Supplier:  Cardinal Health
undefined
Disposable Tissue Grinder System, Covidien
Catalog # 15704-126
Supplier:  Cardinal Health
Supplier Number:  3505SA
Specifications
  • Description:
    Disposable Tissue Grinder System
  • Cat. No.:
    15704-126
  • Supplier no.:
    3505SA
Specifications
About this item
Disposable grinders come ready to use in sterile peel pouch
Complete with 15mL tube.
Grinding is faster and more consistent due to new grinding surface each time. Specimen can be cooled during grinding. Grinder tube doubles as specimen container for transportation and storage. Anti-aerosol sleeve prevents dangerous aerosolization of specimen. Rigid plastic does not break.