Order Entry
Puerto Rico
ContactUsLinkComponent
Whatman™ Anotop Syringe Filters, Non Sterile
Whatman™ Anotop Syringe Filters, Non Sterile
Supplier:  Cytiva
Certificates
undefined
Whatman™ Anotop Syringe Filters, Non Sterile
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • 28138-001
  • 28138-003
  • 28138-005
  • 28138-013
  • 28138-015
  • 28138-017
  • 28138-019
  • Type:
  • Anotop 10 mm syringe filter
  • Anotop 10 mm syringe filter
  • Anotop 10 mm syringe filter
  • Anotop 25 mm syringe filter
  • Anotop 25 mm syringe filter
  • Anotop 25 mm syringe filter
  • Anotop 25 mm syringe filter
  • Dimensions:
  • 15.4×18.5 mm
  • 15.4×18.5 mm
  • 15.4×18.5 mm
  • 36.8×26.3 mm
  • 36.8×26.3 mm
  • 36.8×26.3 mm
  • 36.8×26.3 mm
  • Hold-up Volume:
  • <20 µl
  • <20 µl
  • <20 µl
  • <150 µl
  • <150 µl
  • <150 µl
  • <150 µl
  • Max. Operating Pressure:
  • 6.9 bar (100 psi)
  • 6.9 bar (100 psi)
  • 6.9 bar (100 psi)
  • 6.9 bar (100 psi)
  • 6.9 bar (100 psi)
  • 6.9 bar (100 psi)
  • 6.9 bar (100 psi)

Specifications