Glass Fiber Filters, Extra Thick, Cytiva (Formerly Pall Lab)
: Cytiva
High contaminant-holding capacity makes these filters efficient as depth filters and useful for prefiltration of viscous biological solutions.
- High capacity
- Useful for prefiltration
- Thicker than type A/E
Three times thicker than type A/E filters, with greater contaminant holding capacity. Useful for prefiltration of viscous biological solutions. Contain 5% acrylic binder for additional wet strength.
- Catalog No:
- 28150-735
- 28150-713
- 28150-790
- 28150-779
- Particle Retention:
- 1.0 µm
- 1.0 µm
- 1.0 µm
- 1.0 µm
- Flow Rate:
- Medium-Fast
- Medium-Fast
- Medium-Fast
- Medium-Fast