Avanti™ J-E High-Performance Centrifuge, Beckman Coulter®
Supplier: Avantor
Certificates
About this item
This compact, energy-efficient centrifuge delivers performance while saving valuable laboratory space. Ideal for cellular, genetic, or protein analysis.
- 208/240 V model
- Powerful, High-Torque Switched Reluctance (SR) Drive
- Two Acceleration and Three Deceleration Rates
- Maintenance-Free Partial Vacuum System
- Ideal for Cellular, Genetic, or Protein Analysis
- Centrifuge Compatible with an Extensive Line of Labware
The centrifuge accommodates a variety of rotors including general-purpose, high-capacity, and BioSafety™ certified rotors with patented dual-locking lids for safe sample transportation.
Centrifuge is compatible with an extensive line of labware, including up to twenty-four 96-well microtiter plate processing, four 1 L bottles, and HarvestLine® System liners, which extend the versatility of this centrifuge. Swinging bucket rotors spin volumes of 15, 50, 250, and 500 ml for effective blood component isolation with a variety of gradient kits.
Ordering information: Includes manufacturer’s one-year warranty and a two-year drive-motor warranty. Rotors and optional accessories sold separately. For high-performance centrifuge bottles and tubes, contact your Avantor sales representative.
Caution: 208/240 V model.