ContactUsLinkComponent
BioReady Gold Colloid Passive Conjugation Kit
BioReady Gold Colloid Passive Conjugation Kit
  77680-436
New Product
 :  
BioReady Gold Colloid Passive Conjugation Kit
  77680-436
 :  PCKR
New Product
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Passive conjugation kit for bioready gold colloid
  • Cat. No.:
    77680-436
Use the BioReady passive conjugation kit to conjugate any proteins to 40 nm and 80 nm bare citrate OD 20 gold particles.
Our conjugation kit provides the flexibility to conjugate at any OD from 1 to 20.
 : Kit compenents: BioReady 40 nm bare citrate gold, OD 20, 5 ml, bioready 80 nm bare citrate gold, OD 20, 5 ml, titration buffers covering pH 6 - 9.7, 1 ml each, blocking buffer, 5 ml, conjugate diluent, 5 ml, 10% NaCl for salt test, 5 ml