ContactUsLinkComponent
undefined | Immunological Assays
Catalog # 77680-426
Supplier:  NANOCOMPOSIX, INC.
New Product
CAS Number:  
Catalog # 77680-426
Supplier:  NANOCOMPOSIX, INC.
Supplier Number:  MSKR
New Product
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Material starter kit for lateral flow
  • Cat. No.:
    77680-426
Specifications
About this item
Quickly screen different raw materials to build and optimize your lateral flow assay. Our material starter kit can save you time and money from having to contact multiple suppliers, waiting for long lead times, or purchasing large MOQ.
The product is suitable for developers and researchers interested in sampling from our list of preselected membranes, sample pads, and conjugate pads to give you the best chance of success when optimizing your lateral flow assays.
For quick screening of just membranes, select the option for our Nitrocellulose membranes bundle, which provides 5 different grades of membrane for evaluation.
Delivery: Kit includes: Nitrocellulose membranes, conjugate pads, sample pads, including blood separation pads, wick pads, backing cards, desiccants, storage pouches