VWR® Ham's F-12 Basal Media, Cell Culture Media, Liquid
Catalog # 77661-336
Supplier: VWR International
New Product
CAS Number:
Specifications
- Description:Ham's F12: with 1.0 mM L-Glutamine
- Size:500 ml
- Storage Temperature:2...8 °C
- Cat. No.:77661-336
Specifications
About this item
Ham's F-12 media was originally derived by modifying Ham's F-10 to include increased concentrations of choline, inositol, putrescine, and several amino acids.
- Hydrated using WFI quality water
- 0.1 μm sterile filtered
- Manufactured using ISO 9001:2000 certified processes
It was developed specifically for serum free culturing of Chinese Hamster Ovary (CHO) cells and has also been used with supplementation to grow chondrocytes, primary rat hepatocytes, rat prostate epithelial cells, carcinoma cells, rat skeletal myoblasts and rat, rabbit, and chicken embryos.
Ordering information: Liquid media are supplied in durable, square PETG bottles.