HardyDisks™ Hippurate Disks
Catalog # 77691-386
Supplier: Hardy Diagnostics
New Product
CAS Number:
Specifications
- Pack type:25 disks/jar
- Description:Hippurate disks
- Cat. No.:77691-386
Specifications
About this item
Hippurate disks are used for the sodium hippurate hydrolysis test. Glycine, an end product of hippurate hydrolysis, reacts with ninhydrin to produce a purple color. Hippurate is commonly used to presumptively identify Gardnerella vaginalis, Listeria monocytogenes, Campylobacter jejuni, and group B streptococci.
Product is light, temperature, and moisture sensitive.