Specifications
- Description:Port cap system, with 20 L carboy, 70 mm Port cap, secondary container
- Cat. No.:77677-244
Specifications
About this item
Port cap system, 20 L Nalgene carboy, 70 mm 10- Port cap, secondary container.
Port caps permit multiple waste lines of varying size and type to be connected to the container of your choice. These caps are designed to ensure proper delivery and waste containment of industrial chemicals and solvents. Our closed system reduces harmful emissions which are potentially hazardous to employees and our environment.
Port caps feature ¹/₄ - 28 threaded ports in a PTFE insert, with expanded PTFE gaskets. Each port has a through-hole for ¹/₈" OD tubing, machined at a 10° angle; push tubing through the cap to supply your system. Cap outer is made of autoclavable PP and PTFE.
Port cap features:
Includes plugs for unused holes
Free-spinning polypropylene collar
PTFE inner manifold
(6) ¹/₄-28 UNF ports
(1) ¹/₂" NPT ports
(3) ¹/₄" NPT ports
Carboy features:
Space saving rectangular carboy constructed of HDPE with polyethylene screw closure. Sturdy handle attached to molded in shoulder loops. Molded-in graduations of 1 gallon and 5 L. Increments. Wide mouth for easy filling. 399H×218W×318L mm