Order Entry
Puerto Rico
ContactUsLinkComponent
Port Cap System with 10 L Carboy
Port Cap System with 10 L Carboy
  77677-242
New Product
undefined
Port Cap System with 10 L Carboy
  77677-242
 :  WF-10LP-83-SYS
New Product

 

  • Description:
    Port cap system, 10 L Carboy, 83 B Port cap, secondary container
  • Cat. No.:
    77677-242

 

 

Port cap system, 10 L Carboy, 83 B Port cap, secondary container.

Port caps permit multiple waste lines of varying size and type to be connected to the container of your choice. These caps are designed to ensure proper delivery and waste containment of industrial chemicals and solvents. Our closed system reduces harmful emissions which are potentially hazardous to employees and our environment.

Port caps feature 1/4 - 28 threaded ports in a PTFE insert, with expanded PTFE gaskets. Each port has a through-hole for 1/8" OD tubing, machined at a 10° angle; push tubing through the cap to supply your system. Cap outer is made of autoclavable PP and PTFE.

Port cap features:
83 B Manifold cap may be black or white
Large-bore insert manifold
(1) 1/2" NPT holes
(3) 1/4" NPT holes
(6) 1/4 - 28 holes
Polypropylene with PTFE insert
Includes plugs for unused holes

Carboy features:
Nalgene LDPE Round carboys are a good choice for storage and transport of reagents
Molded-in shoulder loops and graduations in 1 gal. and 5 L increments
Dimensions: 250×389 mm (9.88×15.31")
Molded-in graduations in 5 L and 1 gal. increments; graduated 'To Contain'
Leakproof, unlined white polypropylene closure

 : 10 L Nalgene LDPE Carboy EC-20010, Port Cap Model WF-83B-Kit (includes plugs for all holes, but no tubing adapters. Order adapters based on your tubing size.) Secondary Container SC-83B.