Order Entry
Puerto Rico
ContactUsLinkComponent
IsoReverse Transcriptase
IsoReverse Transcriptase
New Product
Certificates
IsoReverse Transcriptase
New Product

Specifications

  • Storage Conditions:
    −10...−25 °C
  • Shipping Conditions:
    Dry Ice

Specifications

Frequently Bought Together