About this item
Chemically modified Taq DNA polymerase designed for reducing non-specific DNA amplification due to primer-dimer formation. FlashTaq™ is completely suppressed prior to activation and regains full enzymatic activity in only 2 min at 95 °C. Taq DNA Polymerase gene is isolated from Thermus aquaticus YT1 and expressed in E. coli. FlashTaq™ HotStart DNA Polymerase is supplied in a storage buffer containing: HEPES, KCl, EDTA, DTT, Glycerol, and other stabilizers.
- Storage and handling: Upon arrival store at −20 °C for provided expiration date
10X Reaction (MgCl₂) buffer: Specially optimized reaction buffer containing: Tris-HCl, KCl, and MgCl₂. This Buffer is supplied in a 10X concentration and should be diluted for use.
10X Reaction (No MgCl₂) buffer: Specially optimized reaction buffer containing: Tris-HCl (pH 8.4) and KCl. This Buffer is supplied in a 10X concentration and should be diluted for use.
25 mM MgCl₂: 25 mM magnesium chloride.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Environmentally Preferable: