UltraPure Deoxynucleotide Solution Mix
Supplier: EMPIRICAL BIOSCIENCE
New Product
Certificates
About this item
Deoxynucleotide solution mix: A 10 mM equimolar solution of ultrapure dATP, dCTP, dTTP, and dGTP supplied as clear aqueous solution.
Applications: Endpoint PCR, Fast PCR, Fast qPCR, high fidelity PCR, isothermal amplification, reverse transcription PCR/qPCR.
Storage and handling: Upon arrival store at −20 °C for provided expiration date.
Certifications: ISO 13485.
Frequently Bought Together