rEVAlution 2X qPCR MasterMix with Separate ROX
Supplier: EMPIRICAL BIOSCIENCE
New Product
Certificates
About this item
The qPCR MasterMix is supplied as a 2X product and is a ready to use HotStart qPCR MasterMix with EvaGreen® dye, FlashTaq DNA polymerase, dCTP, dGTP, dATP, dTTP, and MgCl2. EvaGreen® is a dsDNA-binding dye. EvaGreen® possesses similar absorption and emission spectra as SYBR® Green I.
- It is cell membrane impermeable
- Non-cytotoxic
- Non-mutagenic
- Safe for aquatic life
FlashTaq is chemically modified hot-start Taq DNA polymerase with an activation time of two minutes at 95 °C. Taq DNA Polymerase gene is isolated from Thermus aquaticus YT1 and expressed in E. coli.
Minimize number of freeze thaw cycles of frozen product.
Applications: qPCR (dye).
Certifications: ISO 13485.
Specifications
- Storage Conditions:Long term storage at –20 °C
Specifications
Frequently Bought Together