Order Entry
Puerto Rico
ContactUsLinkComponent
MagBinder® Fit24 Nucleic Acid Purification System
MagBinder® Fit24 Nucleic Acid Purification System
Catalog # 77749-908
Supplier:  Omega Bio-Tek
New Product
undefined
MagBinder® Fit24 Nucleic Acid Purification System
Catalog # 77749-908
Supplier:  Omega Bio-Tek
Supplier Number:  B1-001-24
New Product

Specifications

  • Description:
    Nucleic acid purification system
  • Method / Format:
    Magnetic beads
  • Display:
    7" touch screen
  • Processing volume:
    50 - 10000 µl
  • W×D×H:
    400×530×480 mm
  • Capacity (Samples/Run):
    1 - 24 samples
  • Cat. No.:
    77749-908

Specifications

About this item

Elevate your laboratory experience with the MagBinder® Fit24 Nucleic acid purification system. Efficiency, affordability, precision, and reliability, all in one powerful instrument.

  • Ease-of-use – Automation-ready cartridges pre-filled with reagents
  • Convenience – No reagent preparation and buffer dispensing
  • High-quality purification – Up to 4 ml samples lysed offline eluted in as little as 50 μl

Simple and efficient – Experience efficient nucleic acid purification with the MagBinder® Fit24 Nucleic acid purification system. This affordable, state-of-the-art instrument utilises 24 magnetic rods coordinated to pick-up, transfer, and release magnetic particles within reagent cartridge wells, ensuring a reliable and streamlined process for DNA and RNA purification. Accommodating 1 to 24 samples simultaneously, the MagBinder® Fit24 is compatible with a range of sample types.

Turnkey solution – Get to processing samples faster with seamless integration of specially designed MB Fit24™ cartridges. The MagBinder® Fit24 comes complete with validated, pre-loaded scripts for MB Fit24™ cartridges, allowing faster set-up time and less manual intervention.

Flexibility at its finest – Designed to work seamlessly with Omega Bio-tek Mag-Bind® kits, the MagBinder® Fit24 presents a flexible, programmable platform. The user-friendly interface empowers effortless programming on new protocols directly on the instrument.