Mouse Recombinant IL-31 (from E. coli)
New Product
Interleukin 31 (IL-31) is an immunoregulatory cytokine that is expressed by activated type 2 T helper (Th2) cells. IL-31 signals through a heterodimer receptor consisting of the IL-31 Receptor A (IL-31RA) and the oncostatin M receptor (OSMR), which are expressed on monocytes, epithelial cells, and keratinocytes. IL-31 promotes allergic reactions and inflammatory skin diseases.
: Ships at room temperature; Store at –20 °C upon delivery.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q6EAL8
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein/Peptide Name:RM IL-31 (AF)
- Purity:≥95%
- Molecular Weight:15.7 kDa
- Sequence:MTCSLSFGAP ISKEDLRTTI DLLKQESQDL YNNYSIKQAS GMSADESIQL PCFSLDREAL TNISVIIAHL EKVKVLSENT VDTSWVIRWL TNISCFNPLN LNISVPGNTD ESYDCKVFVL TVLKQFSNCM AELQAKDNTT C
- Endotoxin Level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Frequently Bought Together