
Paraffin wax blended with synthetic polymers, Solidification point: 56-57°C (132.8-134.6°F), pellets, Tissue-Tek® Paraform® processing/embedding medium Formula 3
Catalog # 89199-380
Supplier: Sakura Finetek
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:8×1 kg
- Cat. No.:89199-380
- Supplier no.:7052
Specifications
Specification Test Results
- Medium Composition:Purified paraffin and synthetic resin blend
- Applications:Processing/Embedding
- Additives:None
- Melting Point:56° to 58°C / 132° to 136°F
- High Temperature Stability:65°C
- Solid Product Range:Pellets
- Sectioning Range:Down to 2µ
Specification Test Results
About this item
:
:GHS07
:1
CAS Number:8002-74-2