Human Recombinant R-Spondin-1 (from CHO cells)
R-Spondin-1 (RSPO-1) is a secreted growth factor that stabilizes β-catenin and enhances WNT signaling.RSPO-1 is expressed in many tissues and regulates female sex determination, XX gonad development, oocyte differentiation, and hematopoietic stem cell specification. The RSPO-1 stabilization of β-catenin promotes intestinal crypt cell proliferation. This suggests that RSPO-1 functions as a stem cell growth factor and may have therapeutic potential in the treatment of gastrointestinal diseases and chemotherapy-induced intestinal mucositis. Additionally, recombinant RSPO-1 protein is used to initiate and maintain intestinal organoid cultures.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity Assay: Luciferase Induction in HEK-293 STF cells in the presence of Murine Wnt-3a, ED₅₀ ≤50 ng/ml, Bioactivity ≥2.0×10⁴ units/mg
- Gene ID:Q2MKA7
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:Roof plate-specific Spondin-1|hRspo1
- Protein/Peptide Name:R-Spondin-1
- Purity:≥95%
- Molecular Weight:26.8 kDa
- Sequence:SRGIKGKRQR RISAEGSQAC AKGCELCSEV NGCLKCSPKL FILLERNDIR QVGVCLPSCP PGYFDARNPD MNKCIKCKIE HCEACFSHNF CTKCKEGLYL HKGRCYPACP EGSSAANGTM ECSSPAQCEM SEWSPWGPCS KKQQLCGFRR GSEERTRRVL HAPVGDHAAC SDTKETRRCT VRRVPCPEGQ KRRKGGQGRR ENANRNLARK ESKEAGAGSR RRKGQQQQQQ QGTVGPLTSA GPA
- Endotoxin Level:Low
- Formulation:Phosphate buffered saline (PBS)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay