Hepatitis C virus (HCV) Recombinant NS3 immunodominant regions
77682-472
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:
- Species:Hepatitis C virus (HCV)
- Environmentally Preferable:
- Protein Synonyms:NS3P|NS3|nonstructural antigen 3|p70|Serine protease/NTPase/helicase|Non structural protein 3
- Protein/Peptide Name:NS3 immunodominant regions
- Purity:HCV NS3 protein is >95% pure as determined by 10% page (coomassie staining)
- Molecular Weight:22 kDa
- Endotoxin Level:Very low
- Formulation:20 mM tris-HCl pH-8, 8 M urea and amp 10 mM B-ME
- Shipping Temperature:Dry Ice
- Cat. No.:77682-472
Recombinant HCV Genotype 1b NS3 Antigen, b-galactosidase-tagged.