Influenza A Recombinant Hemagglutinin
Catalog # 77682-360
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:Influenza A
- Protein Synonyms:Influenza A virus|Influenza virus type A|Influenza A virus A/Uruguay/716/2007(H3N2) Hemagglutinin (propiolactone inactivated)
- Protein/Peptide Name:Hemagglutinin
- Endotoxin Level:Very low
- Shipping Temperature:Dry Ice
- Cat. No.:77682-360
Specifications
About this item
Recombinant Influenza A virus A/Uruguay/716/2007(H3N2) Hemagglutinin Antigen (propiolactone inactivated).