Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
DC-ATADCV

DC-ATADCV

Vial
Catalog # 77682-922
Supplier:   Creative Biomart
Supplier #:  UUM-342
DC-ATADCV
Catalog # 77682-922
Supplier:  Creative Biomart
CAS Number:  
DC-ATADCV
DC-ATADCV

Vial

CAS Number:  
Price
$1,022.60
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

AV-COVID-19.

Specifications

Specifications

  • Pack type:
    Vial
  • Cat. No.:
    77682-922
  • Shipping Temperature:
    Dry Ice
  • Endotoxin Level:
    Very low
  • Protein Synonyms:
    DC-ATADCV
  • Protein/Peptide Name:
    DC-ATADCV

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.