Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Treponema Recombinant TmpA (from E. coli)

Treponema Recombinant TmpA (from E. coli)

Vial
Catalog # 77682-892
Supplier:   Creative Biomart
Supplier #:  TMP-440
Treponema Recombinant TmpA (from E. coli)
Catalog # 77682-892
Supplier:  Creative Biomart
CAS Number:  
Treponema Recombinant TmpA (from E. coli)
Treponema Recombinant TmpA (from E. coli)

Vial

CAS Number:  
Price
$1,022.60
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recombinant Treponema TmpA antigen, his-tagged.

Specifications

Specifications

  • Pack type:
    Vial
  • Cat. No.:
    77682-892
  • Molecular Weight:
    45 kDa
  • Shipping Temperature:
    Dry Ice
  • Protein/Peptide Type:
    Recombinant
  • Formulation:
    Lyophilized from 1 mg/ml in 20 mM sodium carbonate pH-10.
  • Endotoxin Level:
    Very low
  • Purity:
    Greater than 95.0% as determined by SDS-PAGE.
  • Protein Synonyms:
    TmpA|tmpA|Membrane protein A|Antigen tmpA
  • Protein/Peptide Name:
    TmpA
  • Species:
    Treponema
  • Source:
    E. coli

Specifications