TBEV Recombinant glycoprotein E (from E. coli)
Vial
Catalog # 77682-864
Supplier: Creative Biomart
Supplier #: TBE-067
TBEV Recombinant glycoprotein E (from E. coli)
Catalog # 77682-864
Supplier: Creative Biomart
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Recombinant Tick-Borne Encephalitis Virus gE antigen.
Specifications
Specifications
- Pack type:Vial
- Cat. No.:77682-864
- Shipping Temperature:Dry Ice
- Protein/Peptide Type:Recombinant
- Formulation:20 mM Tris-MES pH 6.5, 8 M urea, 200 mM NaCl and 0.05% Tween-20.
- Endotoxin Level:Very low
- Purity:Protein is >90% pure as determined by SDS-PAGE.
- Protein Synonyms:gE|glycoprotein E
- Protein/Peptide Name:glycoprotein E
- Species:TBEV
- Source:E. coli
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



