SARS Recombinant surface glycoprotein (from HEK293)
77682-842
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS
- Protein Synonyms:structural protein|Spike glycoprotein|surface glycoprotein|spike protein|E2|S|spike glycoprotein|Human coronavirus spike glycoprotein|S glycoprotein|E2 glycoprotein
- Protein/Peptide Name:surface glycoprotein
- Purity:Protein is >85% pure as determined SDS-PAGE.
- Endotoxin Level:Very low
- Formulation:SARS CoV-Spike S1 protein solution is supplied in DPBS.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-842
Recombinant SARS Spike Protein, his-tagged.