SARS Recombinant surface glycoprotein (from E. coli)
Catalog # 77682-840
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:SARS
- Protein Synonyms:structural protein|Spike glycoprotein|surface glycoprotein|spike protein|E2|S|spike glycoprotein|Human coronavirus spike glycoprotein|S glycoprotein|E2 glycoprotein
- Protein/Peptide Name:surface glycoprotein
- Purity:Protein is >90% pure as determined SDS-PAGE.
- Endotoxin Level:Very low
- Formulation:SARS Spike protein solution is supplied in PBS.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-840
Specifications
About this item
Recombinant SARS Spike Protein, his-tagged.