SARS Recombinant nucleocapsid phosphoprotein (from HEK293)
Catalog # 77682-836
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS
- Protein Synonyms:Nucleoprotein|structural protein|nucleocapsid phosphoprotein|2019-nCoV nucleocapsid protein|N|NC|ORF9|Protein N
- Protein/Peptide Name:nucleocapsid phosphoprotein
- Purity:Protein is >85% pure as determined SDS-PAGE.
- Endotoxin Level:Very low
- Formulation:SARS Nucleocapsid protein solution is supplied in0.2M NaCl and amp; 50 mM HEPES pH-8.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-836
Specifications
About this item
Recombinant SARS Nucleocaspid Protein, his-tagged.