Order Entry
Puerto Rico
ContactUsLinkComponent
SARS Recombinant nucleocapsid phosphoprotein (from E. coli)
SARS Recombinant nucleocapsid phosphoprotein (from E. coli)
Catalog # 77682-834
Supplier:  Creative Biomart
New Product
CAS Number:  
undefined
SARS Recombinant nucleocapsid phosphoprotein (from E. coli)
Catalog # 77682-834
Supplier:  Creative Biomart
Supplier Number:  SARS-405
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    SARS
  • Protein Synonyms:
    Nucleoprotein|structural protein|nucleocapsid phosphoprotein|2019-nCoV nucleocapsid protein|N|NC|ORF9|Protein N
  • Protein/Peptide Name:
    nucleocapsid phosphoprotein
  • Purity:
    Protein is >90% pure as determined SDS-PAGE.
  • Endotoxin Level:
    Very low
  • Formulation:
    SARS Nucleocaspid protein solution is supplied in PBS.
  • Shipping Temperature:
    Dry Ice
  • Cat. No.:
    77682-834

Specifications

About this item

Recombinant SARS Nucleocaspid Protein.