SARS Recombinant nucleocapsid phosphoprotein
Catalog # 77682-830
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:SARS
- Protein Synonyms:Nucleoprotein|structural protein|nucleocapsid phosphoprotein|2019-nCoV nucleocapsid protein|N|NC|ORF9|Protein N
- Protein/Peptide Name:nucleocapsid phosphoprotein
- Purity:SARS Core protein is >95% pure as determined by 10% PAGE (coomassie staining).
- Endotoxin Level:Very low
- Formulation:50 mM Tris-HCl, 60 mM NaCl and 50% glycerol.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-830
Specifications
About this item
Recombinant SARS Nucleocaspid Protein, GST-tagged.